6-Chlor-2-fluor-3-methylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

526622-5G
SIAL526622-5GEA 87 EUR
SIAL526622-5G
6-Chlor-2-fluor-3-methylbenzaldehyd, Sigma-Aldrich®
6-Chlor-2-fluor-3-methylbenzaldehyd
Formel: C₈H₆ClFO
Molecular Weight: 172,59 g/mol
MDL: MFCD01631370
CAS-Nummer: 286474-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR