Cyclopropylmagnesiumbromid 0.5 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

526797-100ML
SIAL526797-100MLEA 335 EUR
SIAL526797-100ML
Cyclopropylmagnesiumbromid 0.5 mol/l in THF, Sigma-Aldrich®
Cyclopropylmagnesiumbromid
Formel: C₃H₅BrMg
Molecular Weight: 145,28 g/mol
MDL: MFCD00672094
CAS-Nummer: 23719-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR