3-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

M-Clarity™ Qualitätsstufe = MQ100

527548-1G
SIAL527548-1GEA 34 EUR
SIAL527548-1G
3-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
MDL: MFCD03093896
CAS-Nummer: 269410-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR