4-Ethylbenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Ethylbenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

527726-1ML
SIAL527726-1MLEA 58.4 EUR
SIAL527726-1ML
4-Ethylbenzonitril, Sigma-Aldrich®
4-Ethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00016383
CAS-Nummer: 25309-65-3
EINECS: 246-811-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR