4-(Hydroxymethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Cyanobenzyl alcohol

527750-10G
SIAL527750-10GEA 442 EUR
SIAL527750-10G
4-(Hydroxymethyl)benzonitril, Sigma-Aldrich®
4-(Hydroxymethyl)benzonitril
Formel: C₈H₇NO
MDL: MFCD00870633
CAS-Nummer: 874-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR