4-Phenoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

527769-1G
SIAL527769-1GEA 57.1 EUR
SIAL527769-1G
4-Phenoxybenzonitril, Sigma-Aldrich®
4-Phenoxybenzonitril
Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
MDL: MFCD00017346
CAS-Nummer: 3096-81-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR