4-Butylbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

527777-5G
SIAL527777-5GEA 102 EUR
SIAL527777-5G
4-Butylbenzonitril, Sigma-Aldrich®
4-Butylbenzonitril
Formel: C₁₁H₁₃N
Molecular Weight: 159,23 g/mol
MDL: MFCD00052345
CAS-Nummer: 20651-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR