(R,R)-(-)-2,5-Hexandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R,R)-(-)-2,5-Dihydroxyhexan

52792-1G
SIAL52792-1GEA 129 EUR
SIAL52792-1G
(R,R)-(-)-2,5-Hexandiol, Sigma-Aldrich®
(R,R)-(-)-2,5-Hexandiol
Formel: C₆H₁₄O₂
Molecular Weight: 118,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00142339
CAS-Nummer: 17299-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR