N-Chlorphthalimid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

528218-25G
SIAL528218-25GEA 97.2 EUR
SIAL528218-25G
N-Chlorphthalimid, Sigma-Aldrich®
N-Chlorphthalimid
Formel: C₈H₄ClNO₂
MDL: MFCD00023027
CAS-Nummer: 3481-09-2
EINECS: 222-459-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR