3-Ethoxybenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

528234-5ML
SIAL528234-5MLEA 114 EUR
SIAL528234-5ML
3-Ethoxybenzylalkohol, Sigma-Aldrich®
3-Ethoxybenzylalkohol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00016864
CAS-Nummer: 71648-21-0
EINECS: 275-749-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR