2-Methoxy-1-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

528382-1G
SIAL528382-1GEA 134 EUR
SIAL528382-1G
2-Methoxy-1-naphthoesäure, Sigma-Aldrich®
2-Methoxy-1-naphthoesäure
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,206 g/mol
MDL: MFCD00021532
CAS-Nummer: 947-62-6
EINECS: 213-429-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR