3-Ethoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

528498-10G
SIAL528498-10GEA 160 EUR
SIAL528498-10G
3-Ethoxybenzoesäure, Sigma-Aldrich®
3-Ethoxybenzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00016517
CAS-Nummer: 621-51-2
EINECS: 210-690-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR