3-Methoxy-2-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methoxynaphthalene-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

528544-1G
SIAL528544-1GEA 189 EUR
SIAL528544-1G
3-Methoxy-2-naphthoesäure, Sigma-Aldrich®
3-Methoxy-2-naphthoesäure
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,2060 g/mol
MDL: MFCD00094168
CAS-Nummer: 883-62-5
EINECS: 212-932-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR