Methyl-3-methoxy-4-methylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-methoxy-4-methylbenzoate

M-Clarity™ Qualitätsstufe = MQ100

528781-25G
SIAL528781-25GEA 766 EUR
SIAL528781-25G
Methyl-3-methoxy-4-methylbenzoat, Sigma-Aldrich®
Methyl-3-methoxy-4-methylbenzoat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,21 g/mol
MDL: MFCD00082710
CAS-Nummer: 3556-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR