Methyl-2-amino-5-brombenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-amino-5-bromobenzoate

M-Clarity™ Qualitätsstufe = MQ100

528811-25G
SIAL528811-25GEA 90.8 EUR
SIAL528811-25G
Methyl-2-amino-5-brombenzoat, Sigma-Aldrich®
Methyl-2-amino-5-brombenzoat
Formel: C₈H₈BrNO₂
Molecular Weight: 230,06 g/mol
MDL: MFCD00144761
CAS-Nummer: 52727-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR