2-Brom-5-fluoranilin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

528900-25G
SIAL528900-25GEA 41.9 EUR
SIAL528900-25G
2-Brom-5-fluoranilin, Sigma-Aldrich®
2-Brom-5-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190,01 g/mol
MDL: MFCD00070750
CAS-Nummer: 1003-99-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR