4-Fluor-2-methoxyphenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

528943-5G
SIAL528943-5GEA 105 EUR
SIAL528943-5G
4-Fluor-2-methoxyphenol, Sigma-Aldrich®
4-Fluor-2-methoxyphenol
Formel: C₇H₇FO₂
Molecular Weight: 142,13 g/mol
MDL: MFCD00070797
CAS-Nummer: 450-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR