Methyl-2-fluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluorbenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

528951-5G
SIAL528951-5GEA 63.5 EUR
SIAL528951-5G
Methyl-2-fluorbenzoat, Sigma-Aldrich®
Methyl-2-fluorbenzoat
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00017913
CAS-Nummer: 394-35-4
EINECS: 206-894-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR