2-Brom-5-fluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

528978-5G
SIAL528978-5GEA 71.8 EUR
SIAL528978-5G
2-Brom-5-fluorbenzaldehyd, Sigma-Aldrich®
2-Bromo-5-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203.01 g/mol
MDL: MFCD00142872
CAS-Nummer: 94569-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR