2-Fluor-p-anisaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-4-anisaldehyd, 2-Fluor-4-methoxybenzaldehyd

M-Clarity™ Qualitätsstufe = MQ100

528986-25G
SIAL528986-25GEA 168 EUR
SIAL528986-25G
2-Fluor-p-anisaldehyd, Sigma-Aldrich®
2-Fluor-p-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00236679
CAS-Nummer: 331-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR