2,4-Dihydroxy-3-nitro-6-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-6-methyl-3-nitro-2-pyridon, 2,4-Dihydroxy-6-methyl-3-nitropyridin

M-Clarity™ Qualitätsstufe = MQ100

529001-25G
SIAL529001-25GEA 209 EUR
SIAL529001-25G
2,4-Dihydroxy-3-nitro-6-picolin, Sigma-Aldrich®
2,4-Dihydroxy-3-nitro-6-picolin
Formel: C₆H₆N₂O₄
Molecular Weight: 170,12 g/mol
MDL: MFCD00167612
CAS-Nummer: 4966-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR