3-Iodbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

529036-10G 529036-1G
SIAL529036-10GEA 148 EUR
SIAL529036-10G SIAL529036-1G
3-Iodbenzonitril, Sigma-Aldrich®
3-Iodbenzonitril
Formel: C₇H₄IN
Molecular Weight: 229.02 g/mol
MDL: MFCD00079762
CAS-Nummer: 69113-59-3
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR