3-Chlor-5-fluoranisol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Chloro-3-fluoro-5-methoxybenzene

M-Clarity™ Qualitätsstufe = MQ100

529060-10G
SIAL529060-10GEA 222 EUR
SIAL529060-10G
3-Chlor-5-fluoranisol, Sigma-Aldrich®
3-Chlor-5-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD00070770
CAS-Nummer: 202925-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR