2-Chlor-5-iodtoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Chlor-4-iod-2-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

529087-10G
SIAL529087-10GEA 285 EUR
SIAL529087-10G
2-Chlor-5-iodtoluol, Sigma-Aldrich®
2-Chlor-5-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252,48 g/mol
MDL: MFCD03093903
CAS-Nummer: 116632-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR