4-(Trifluormethylthio)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-((Trifluoromethyl)thio)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

529141-5G
SIAL529141-5GEA 91.7 EUR
SIAL529141-5G
4-(Trifluormethylthio)benzaldehyd, Sigma-Aldrich®
4-(Trifluoromethylthio)benzaldehyde
Formel: C₈H₅F₃OS
Molecular Weight: 206,19 g/mol
MDL: MFCD00129191
CAS-Nummer: 4021-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR