2-(Trifluormethoxy)benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
529168-5G
SIAL529168-5GEA
48.7
EUR
SIAL529168-5G
2-(Trifluormethoxy)benzaldehyd, Sigma-Aldrich®
2-(Trifluormethoxy)benzaldehyd
Formel:
C₈H₅F₃O₂ |
MDL:
MFCD00042405 CAS-Nummer: 94651-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...