3-Fluor-5-(trifluormethyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-fluoro-5-(trifluoromethyl)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ200

529206-5G
SIAL529206-5GEA 71.8 EUR
SIAL529206-5G
3-Fluor-5-(trifluormethyl)benzaldehyd, Sigma-Aldrich®
3-Fluoro-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
MDL: MFCD00061312
CAS-Nummer: 188815-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR