Certriacetat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Essigsäure Cer(III)salz Hydrat, Cer(III)acetat Hydrat

M-Clarity™ Qualitätsstufe = MQ100

529559-10G
SIAL529559-10GEA 82.9 EUR
SIAL529559-10G
Certriacetat Hydrat, Sigma-Aldrich®
Certriacetat Hydrat
Formel: C₆H₉CeO₆·H₂O
Molecular Weight: 335,266 g/mol
MDL: MFCD00150533
CAS-Nummer: 206996-60-3
EINECS: 208-654-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR