Bis(triphenylphosphin)nickel(II)chlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

M-Clarity™ Qualitätsstufe = MQ100

529621-10G
SIAL529621-10GEA 132 EUR
SIAL529621-10G
Bis(triphenylphosphin)nickel(II)chlorid, Sigma-Aldrich®
Bis(triphenylphosphin)nickel(II)chlorid
Formel: C₃₆H₃₂Cl₂NiP₂
Molecular Weight: 656,19 g/mol
MDL: MFCD00009592
CAS-Nummer: 14264-16-5
EINECS: 238-154-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR