(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

529907-1G
SIAL529907-1GEA 83.3 EUR
SIAL529907-1G
(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester, Sigma-Aldrich®
(E)-1-Penten-1,2-diboronsäurebis(pinakol)ester
Formel: C₁₇H₃₂B₂O₄
MDL: MFCD03093909
CAS-Nummer: 307531-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR