Diphenyliodoniumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

530972-1G
SIAL530972-1GEA 174 EUR
SIAL530972-1G
Diphenyliodoniumtrifluormethansulfonat, Sigma-Aldrich®
Diphenyliodonium trifluoromethanesulphonate
Formel: C₁₃H₁₀F₃IO₃S
Molecular Weight: 430,19 g/mol
MDL: MFCD00191356
CAS-Nummer: 66003-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR