Ethyl-6-chlornicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 6-chloronicotinate

531197-25G
SIAL531197-25GEA 28.2 EUR
SIAL531197-25G
Ethyl-6-chlornicotinat, Sigma-Aldrich®
Ethyl-6-chlornicotinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD00082739
CAS-Nummer: 49608-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR