Acetaldehyd-[13C2] Isotope_(99% 13C), Sigma-Aldrich®

Lieferant: Merck
Gefahr


MQ200

531227-1G
SIAL531227-1GEA 1330 EUR
SIAL531227-1G
Acetaldehyd-[13C2] Isotope_(99% 13C), Sigma-Aldrich®
Acetaldehyd-[13C2]
Formel: C₂H₄O
Lagertemperatur: Kühlschrank
MDL: MFCD00143529
CAS-Nummer: 1632-98-0
UN: 1089
ADR: 3 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR