2,2-Difluor-2-(fluorsulfonyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

531413-25G
SIAL531413-25GEA 104 EUR
SIAL531413-25G
2,2-Difluor-2-(fluorsulfonyl)essigsäure, Sigma-Aldrich®
2,2-Difluor-2-(fluorsulfonyl)essigsäure
Formel: C₂HF₃O₄S
MDL: MFCD01320807
CAS-Nummer: 1717-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR