Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

531421-25G
SIAL531421-25GEA 176 EUR
SIAL531421-25G
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat, Sigma-Aldrich®
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel: C₅H₉F₃O₄SSi
MDL: MFCD02093343
CAS-Nummer: 120801-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR