Bis(pyridin)iodoniumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
531634-1G
SIAL531634-1GEA
68.1
EUR
SIAL531634-1G
Bis(pyridin)iodoniumtetrafluoroborat, Sigma-Aldrich®
Bis(pyridin)iodoniumtetrafluoroborat
Formel:
C₁₀H₁₀BF₄IN₂ Molecular Weight: 371,91 g/mol |
MDL:
MFCD03703393 CAS-Nummer: 15656-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...