6-Brom-2,4,5-trichlorphenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

531642-1G
SIAL531642-1GEA 134 EUR
SIAL531642-1G
6-Brom-2,4,5-trichlorphenol, Sigma-Aldrich®
6-Brom-2,4,5-trichlorphenol
Formel: C₆H₂BrCl₃O
CAS-Nummer: 4524-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR