2'-Hydroxy-4'-methylacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 1-(2-Hydroxy-4-methylphenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

531669-10G
SIAL531669-10GEA 127 EUR
SIAL531669-10G
2'-Hydroxy-4'-methylacetophenon, Sigma-Aldrich®
2'-Hydroxy-4'-methylacetophenon
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
MDL: MFCD00100559
CAS-Nummer: 6921-64-8
EINECS: 230-040-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR