m-Tolylacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

531677-25G
SIAL531677-25GEA 48.2 EUR
SIAL531677-25G
m-Tolylacetat, Sigma-Aldrich®
m-Tolylacetat
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
MDL: MFCD00041910
CAS-Nummer: 122-46-3
EINECS: 204-546-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR