1-Butyl-3-methylimidazoliummethylsulfat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
53177-100G-F
SIAL53177-100G-FEA
200
EUR
SIAL53177-100G-F
1-Butyl-3-methylimidazoliummethylsulfat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium methyl sulphate
Formel:
C₉H₁₈N₂O₄S Dichte: 1,21 g/cm³ (20 °C) |
MDL:
MFCD03095437 CAS-Nummer: 401788-98-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...