1-Benzyloxy-4-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Benzyloxy)-4-iodobenzene, 4-Benzyloxyiodobenzene

M-Clarity™ Qualitätsstufe = MQ100

532185-5G
SIAL532185-5GEA 127 EUR
SIAL532185-5G
1-Benzyloxy-4-iodbenzol, Sigma-Aldrich®
1-Benzyloxy-4-iodbenzol
Formel: C₁₃H₁₁IO
Molecular Weight: 310,13 g/mol
MDL: MFCD00079704
CAS-Nummer: 19578-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR