1-Benzyloxy-3-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Benzyloxy)-3-iodobenzene

M-Clarity™ Qualitätsstufe = MQ100

532193-10G
SIAL532193-10GEA 228 EUR
SIAL532193-10G
1-Benzyloxy-3-iodbenzol, Sigma-Aldrich®
1-Benzyloxy-3-iodbenzol
Formel: C₁₃H₁₁IO
Molecular Weight: 310,13 g/mol
MDL: MFCD01318100
CAS-Nummer: 107623-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR