Methyl-4-iod-2-methoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Iod-2-methoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

532568-25G
SIAL532568-25GEA 177 EUR
SIAL532568-25G
Methyl-4-iod-2-methoxybenzoat, Sigma-Aldrich®
Methyl-4-iod-2-methoxybenzoat
Formel: C₉H₉IO₃
Molecular Weight: 292,07 g/mol
MDL: MFCD00052925
CAS-Nummer: 35387-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR