Chromon-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

532576-5G
SIAL532576-5GEA 112 EUR
SIAL532576-5G
Chromon-3-carbonsäure, Sigma-Aldrich®
Chromon-3-carbonsäure
Formel: C₁₀H₆O₄
Molecular Weight: 190,15 g/mol
MDL: MFCD00017338
CAS-Nummer: 39079-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR