5-Brom-2-fluorbenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (5-Bromo-2-fluorophenyl)methanamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

532762-25G
SIAL532762-25GEA 552 EUR
SIAL532762-25G
5-Brom-2-fluorbenzylamin Hydrochlorid, Sigma-Aldrich®
5-Brom-2-fluorbenzylamin Hydrochlorid
Formel: C₇H₈BrClFN
Molecular Weight: 240,50 g/mol
MDL: MFCD00143426
CAS-Nummer: 202865-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR