3,3,4,4,5,5,6,6,6-Nonafluor-1-hexanol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
532770-25G
SIAL532770-25GEA
235
EUR
SIAL532770-25G
3,3,4,4,5,5,6,6,6-Nonafluor-1-hexanol, Sigma-Aldrich®
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
Formel:
C₆H₅F₉O |
MDL:
MFCD00039543 CAS-Nummer: 2043-47-2 EINECS: 218-050-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...