4,4,5,5,6,6,7,7,7-Nonafluor-1-heptanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

532797-25G
SIAL532797-25GEA 311 EUR
SIAL532797-25G
4,4,5,5,6,6,7,7,7-Nonafluor-1-heptanol, Sigma-Aldrich®
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol
Formel: C₇H₇F₉O
MDL: MFCD02183552
CAS-Nummer: 83310-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR