Methyl-4-brom-3-methylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-bromo-3-methylbenzoate

M-Clarity™ Qualitätsstufe = MQ100

532878-25G
SIAL532878-25GEA 71.8 EUR
SIAL532878-25G
Methyl-4-brom-3-methylbenzoat, Sigma-Aldrich®
Methyl-4-brom-3-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
MDL: MFCD00673014
CAS-Nummer: 148547-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR