(R)-(-)-3-Chinuclidinol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
532908-5G
SIAL532908-5GEA
201
EUR
SIAL532908-5G
(R)-(-)-3-Chinuclidinol Hydrochlorid, Sigma-Aldrich®
(R)-(-)-3-Chinuclidinol Hydrochlorid
Formel:
C₇H₁₄ClNO Molecular Weight: 163,65 g/mol |
CAS-Nummer:
42437-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...