4-Brom-2,6-difluorbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Bromo-2,6-difluorophenyl)methanol

M-Clarity™ Qualitätsstufe = MQ100

533033-5G
SIAL533033-5GEA 76.9 EUR
SIAL533033-5G
4-Brom-2,6-difluorbenzylalkohol, Sigma-Aldrich®
4-Brom-2,6-difluorbenzylalkohol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD03094461
CAS-Nummer: 162744-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR